Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616929 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTERF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MTERF2 - mitochondrial transcription termination factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033050.2 | Intron | NP_001028222.1 | ||||
NM_025198.4 | Intron | NP_079474.2 | ||||
XM_011538767.2 | Intron | XP_011537069.1 | ||||
XM_017019985.1 | Intron | XP_016875474.1 | ||||
XM_017019986.1 | Intron | XP_016875475.1 | ||||
XM_017019987.1 | Intron | XP_016875476.1 |
TMEM263 - transmembrane protein 263 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |