Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600521 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MASP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MASP1 - mannan binding lectin serine peptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031849.2 | 3128 | Intron | NP_001027019.1 | |||
NM_001879.5 | 3128 | UTR 3 | NP_001870.3 | |||
NM_139125.3 | 3128 | Intron | NP_624302.1 | |||
XM_006713701.2 | 3128 | Intron | XP_006713764.1 | |||
XM_011512989.2 | 3128 | Intron | XP_011511291.1 | |||
XM_011512990.2 | 3128 | Intron | XP_011511292.1 | |||
XM_011512991.2 | 3128 | Intron | XP_011511293.1 | |||
XM_017006869.1 | 3128 | Intron | XP_016862358.1 | |||
XM_017006870.1 | 3128 | Intron | XP_016862359.1 | |||
XM_017006871.1 | 3128 | Intron | XP_016862360.1 | |||
XM_017006872.1 | 3128 | Intron | XP_016862361.1 |
Set Membership: |
HapMap |