Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCTGAGGGACCATCTCTGGGAG[A/G]GAGTCTAGGCCCAGCCTTGGATGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614910 MIM: 182453 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QTNF6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1QTNF6 - C1q and tumor necrosis factor related protein 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SSTR3 - somatostatin receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001051.4 | 2884 | UTR 3 | NP_001042.1 | |||
NM_001278687.2 | 2884 | UTR 3 | NP_001265616.1 | |||
XM_005261721.4 | 2884 | UTR 3 | XP_005261778.1 | |||
XM_006724311.2 | 2884 | UTR 3 | XP_006724374.1 | |||
XM_011530349.2 | 2884 | UTR 3 | XP_011528651.1 | |||
XM_017028923.1 | 2884 | UTR 3 | XP_016884412.1 | |||
XM_017028924.1 | 2884 | UTR 3 | XP_016884413.1 |