Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGTACCCCTGGGTCACCGCACT[C/T]AGCACAGCAGAGCTGTGTGGCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616386 MIM: 609862 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCTD17 - potassium channel tetramerization domain containing 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282684.1 | Intron | NP_001269613.1 | ||||
NM_001282685.1 | Intron | NP_001269614.1 | ||||
NM_001282686.1 | Intron | NP_001269615.1 | ||||
NM_024681.3 | Intron | NP_078957.2 | ||||
XM_005261741.3 | Intron | XP_005261798.2 | ||||
XM_005261742.2 | Intron | XP_005261799.2 | ||||
XM_005261743.2 | Intron | XP_005261800.2 | ||||
XM_005261744.2 | Intron | XP_005261801.1 | ||||
XM_011530374.2 | Intron | XP_011528676.1 | ||||
XM_011530377.2 | Intron | XP_011528679.1 |
TMPRSS6 - transmembrane protease, serine 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |