Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616386 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD17 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KCTD17 - potassium channel tetramerization domain containing 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282684.1 | Intron | NP_001269613.1 | ||||
NM_001282685.1 | Intron | NP_001269614.1 | ||||
NM_001282686.1 | Intron | NP_001269615.1 | ||||
NM_024681.3 | Intron | NP_078957.2 | ||||
XM_005261741.3 | Intron | XP_005261798.2 | ||||
XM_005261742.2 | Intron | XP_005261799.2 | ||||
XM_005261743.2 | Intron | XP_005261800.2 | ||||
XM_005261744.2 | Intron | XP_005261801.1 | ||||
XM_011530374.2 | Intron | XP_011528676.1 | ||||
XM_011530377.2 | Intron | XP_011528679.1 |