Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606542 MIM: 612187 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDAC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HDAC7 - histone deacetylase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098416.3 | 3623 | UTR 3 | NP_001091886.1 | |||
NM_001308090.1 | 3623 | UTR 3 | NP_001295019.1 | |||
NM_015401.4 | 3623 | UTR 3 | NP_056216.2 | |||
XM_011538478.1 | 3623 | UTR 3 | XP_011536780.1 | |||
XM_011538479.1 | 3623 | UTR 3 | XP_011536781.1 | |||
XM_011538480.1 | 3623 | UTR 3 | XP_011536782.1 | |||
XM_011538481.1 | 3623 | UTR 3 | XP_011536783.1 | |||
XM_011538482.1 | 3623 | UTR 3 | XP_011536784.1 | |||
XM_011538483.1 | 3623 | Intron | XP_011536785.1 | |||
XM_017019455.1 | 3623 | UTR 3 | XP_016874944.1 | |||
XM_017019456.1 | 3623 | Intron | XP_016874945.1 |
SLC48A1 - solute carrier family 48 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017842.2 | 3623 | Intron | NP_060312.2 | |||
XM_005269016.4 | 3623 | Intron | XP_005269073.1 | |||
XM_011538558.2 | 3623 | Intron | XP_011536860.1 | |||
XM_017019612.1 | 3623 | Intron | XP_016875101.1 | |||
XM_017019613.1 | 3623 | Intron | XP_016875102.1 | |||
XM_017019614.1 | 3623 | Intron | XP_016875103.1 | |||
XM_017019615.1 | 3623 | Intron | XP_016875104.1 | |||
XM_017019616.1 | 3623 | Intron | XP_016875105.1 | |||
XM_017019617.1 | 3623 | Intron | XP_016875106.1 |
Set Membership: |
Validated |