Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGAGGCCAGAAAGGGGCAGAAC[A/G]GCAAGAGACAGGAAGGTCACTGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607814 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD27 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ANKRD27 - ankyrin repeat domain 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032139.2 | Intron | NP_115515.2 | ||||
XM_006723412.2 | Intron | XP_006723475.1 | ||||
XM_017027349.1 | Intron | XP_016882838.1 |
RGS9BP - regulator of G-protein signaling 9 binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |