Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGTTGTCTCAATCTTGTTGACA[C/T]TCAGGGCTGCTGACATCACTGATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 124070 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP2F1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYP2F1 - cytochrome P450 family 2 subfamily F member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000774.4 | Intron | NP_000765.2 | ||||
XM_011526551.2 | Intron | XP_011524853.1 | ||||
XM_011526552.2 | Intron | XP_011524854.1 | ||||
XM_011526553.2 | Intron | XP_011524855.1 | ||||
XM_011526554.2 | Intron | XP_011524856.1 | ||||
XM_011526555.2 | Intron | XP_011524857.1 | ||||
XM_017026384.1 | Intron | XP_016881873.1 | ||||
XM_017026385.1 | Intron | XP_016881874.1 | ||||
XM_017026386.1 | Intron | XP_016881875.1 | ||||
XM_017026387.1 | Intron | XP_016881876.1 |
Set Membership: |
HapMap |