Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAATTGTTTGCTACTGTTGAGTCC[A/G]GGTGGAACAGGCATATGAGAAAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611995 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MANSC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MANSC4 - MANSC domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146221.1 | 750 | Silent Mutation | CCC,CCT | P,P 250 | NP_001139693.1 | |
XM_006719024.3 | 750 | Silent Mutation | CCC,CCT | P,P 205 | XP_006719087.1 | |
XM_011520542.2 | 750 | Silent Mutation | CCC,CCT | P,P 223 | XP_011518844.1 |
MRPS35 - mitochondrial ribosomal protein S35 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |