Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGCGGCAGTTCCGGGAGGCAGCC[A/G]AAAGTACTAACAGTTGGTAGCAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTBD11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BTBD11 - BTB domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017523.1 | 1625 | Intron | NP_001017523.1 | |||
NM_001018072.1 | 1625 | UTR 5 | NP_001018082.1 | |||
XM_005268645.3 | 1625 | Intron | XP_005268702.1 | |||
XM_011537909.2 | 1625 | Intron | XP_011536211.1 | |||
XM_011537910.2 | 1625 | Intron | XP_011536212.1 | |||
XM_017018807.1 | 1625 | UTR 5 | XP_016874296.1 |
Set Membership: |
HapMap |