Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATAAAGTGAAGCATACCTTATTG[G/T]GATTCTAACTAACTAGGTAAGCCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603246 MIM: 616650 | ||||||||||||||||||||
Literature Links: |
GTF3C1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GTF3C1 - general transcription factor IIIC subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIAA0556 - KIAA0556 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015202.3 | Intron | NP_056017.3 | ||||
XM_005255201.3 | Intron | XP_005255258.1 | ||||
XM_005255202.3 | Intron | XP_005255259.1 | ||||
XM_005255203.3 | Intron | XP_005255260.1 | ||||
XM_005255206.3 | Intron | XP_005255263.1 | ||||
XM_005255208.2 | Intron | XP_005255265.1 | ||||
XM_006721025.3 | Intron | XP_006721088.1 | ||||
XM_011545773.2 | Intron | XP_011544075.1 | ||||
XM_011545774.1 | Intron | XP_011544076.1 | ||||
XM_011545775.2 | Intron | XP_011544077.1 | ||||
XM_011545776.2 | Intron | XP_011544078.1 | ||||
XM_011545777.2 | Intron | XP_011544079.1 | ||||
XM_017023085.1 | Intron | XP_016878574.1 | ||||
XM_017023086.1 | Intron | XP_016878575.1 | ||||
XM_017023087.1 | Intron | XP_016878576.1 | ||||
XM_017023088.1 | Intron | XP_016878577.1 |