Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGTGGATATTTTTCAAAAGTTAT[C/T]TATTATAATATTAGTGGGGTTTTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SIMC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SIMC1 - SUMO interacting motifs containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308195.1 | Intron | NP_001295124.1 | ||||
NM_001308196.1 | Intron | NP_001295125.1 | ||||
NM_001308200.1 | Intron | NP_001295129.1 | ||||
NM_198567.5 | Intron | NP_940969.3 | ||||
XM_011534553.2 | Intron | XP_011532855.1 | ||||
XM_011534554.2 | Intron | XP_011532856.1 | ||||
XM_011534556.2 | Intron | XP_011532858.1 | ||||
XM_017009454.1 | Intron | XP_016864943.1 | ||||
XM_017009455.1 | Intron | XP_016864944.1 | ||||
XM_017009456.1 | Intron | XP_016864945.1 |