Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGGTGTCCAACCAGCCTCTACT[A/G]TGTTTCCATACTGGGACATACAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607315 MIM: 607443 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CABP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CABP5 - calcium binding protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019855.4 | Intron | NP_062829.1 | ||||
XM_017026992.1 | Intron | XP_016882481.1 |
ELSPBP1 - epididymal sperm binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |