Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611697 | ||||||||||||||||||||
Literature Links: |
PSMG4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSMG4 - proteasome assembly chaperone 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128591.1 | 6096 | Intron | NP_001122063.1 | |||
NM_001128592.1 | 6096 | Intron | NP_001122064.1 | |||
NM_001135750.1 | 6096 | Intron | NP_001129222.1 | |||
XM_011514591.2 | 6096 | Intron | XP_011512893.1 | |||
XM_011514593.2 | 6096 | Intron | XP_011512895.1 | |||
XM_011514594.2 | 6096 | Intron | XP_011512896.1 | |||
XM_011514595.2 | 6096 | Intron | XP_011512897.1 |
SLC22A23 - solute carrier family 22 member 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286455.1 | 6096 | UTR 3 | NP_001273384.1 | |||
NM_001286456.1 | 6096 | Intron | NP_001273385.1 | |||
NM_015482.1 | 6096 | UTR 3 | NP_056297.1 | |||
NM_021945.6 | 6096 | UTR 3 | NP_068764.3 | |||
XM_011514801.2 | 6096 | UTR 3 | XP_011513103.1 | |||
XM_017011180.1 | 6096 | UTR 3 | XP_016866669.1 | |||
XM_017011181.1 | 6096 | UTR 3 | XP_016866670.1 | |||
XM_017011182.1 | 6096 | UTR 3 | XP_016866671.1 | |||
XM_017011183.1 | 6096 | Intron | XP_016866672.1 | |||
XM_017011184.1 | 6096 | UTR 3 | XP_016866673.1 | |||
XM_017011185.1 | 6096 | Intron | XP_016866674.1 | |||
XM_017011186.1 | 6096 | UTR 3 | XP_016866675.1 |