Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300642 MIM: 300723 | ||||||||||||||||||||||||||||||||
Literature Links: |
SRPX2 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
SRPX2 - sushi repeat containing protein, X-linked 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYTL4 - synaptotagmin like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129896.2 | 3940 | UTR 3 | NP_001123368.1 | |||
NM_001174068.1 | 3940 | UTR 3 | NP_001167539.1 | |||
NM_080737.2 | 3940 | UTR 3 | NP_542775.2 | |||
XM_005262229.2 | 3940 | UTR 3 | XP_005262286.1 | |||
XM_011531068.2 | 3940 | UTR 3 | XP_011529370.1 | |||
XM_017029966.1 | 3940 | UTR 3 | XP_016885455.1 | |||
XM_017029967.1 | 3940 | UTR 3 | XP_016885456.1 | |||
XM_017029968.1 | 3940 | UTR 3 | XP_016885457.1 | |||
XM_017029969.1 | 3940 | UTR 3 | XP_016885458.1 | |||
XM_017029970.1 | 3940 | UTR 3 | XP_016885459.1 | |||
XM_017029971.1 | 3940 | UTR 3 | XP_016885460.1 |