Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606804 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GOLM1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GOLM1 - golgi membrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016548.3 | 2314 | UTR 3 | NP_057632.2 | |||
NM_177937.2 | 2314 | UTR 3 | NP_808800.1 |
NAA35 - N(alpha)-acetyltransferase 35, NatC auxiliary subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321881.1 | 2314 | Intron | NP_001308810.1 | |||
NM_001321882.1 | 2314 | Intron | NP_001308811.1 | |||
NM_024635.3 | 2314 | Intron | NP_078911.3 | |||
XM_005252127.3 | 2314 | Intron | XP_005252184.1 | |||
XM_011518903.2 | 2314 | Intron | XP_011517205.1 | |||
XM_011518904.2 | 2314 | Intron | XP_011517206.1 | |||
XM_017015012.1 | 2314 | Intron | XP_016870501.1 |
Set Membership: |
HapMap |