Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGCACTGCAGCAAGAAGGCTGA[C/T]GCTCAGTATGTGGAACGTCTGGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610070 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102724776 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC102724776 - uncharacterized LOC102724776 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAP9 - microtubule associated protein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039580.1 | 3851 | UTR 3 | NP_001034669.1 | |||
XM_006714306.3 | 3851 | UTR 3 | XP_006714369.1 | |||
XM_011532253.1 | 3851 | UTR 3 | XP_011530555.1 | |||
XM_011532254.2 | 3851 | UTR 3 | XP_011530556.1 | |||
XM_011532255.2 | 3851 | UTR 3 | XP_011530557.1 | |||
XM_017008616.1 | 3851 | UTR 3 | XP_016864105.1 | |||
XM_017008617.1 | 3851 | UTR 3 | XP_016864106.1 | |||
XM_017008618.1 | 3851 | Intron | XP_016864107.1 |