Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602352 MIM: 611988 MIM: 176884 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNRH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GNRH2 - gonadotropin releasing hormone 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001310220.1 | 313 | Intron | NP_001297149.1 | |||
NM_001501.1 | 313 | Intron | NP_001492.1 | |||
NM_178331.1 | 313 | Intron | NP_847901.1 | |||
NM_178332.1 | 313 | Intron | NP_847902.1 | |||
XM_017027823.1 | 313 | Intron | XP_016883312.1 |
MRPS26 - mitochondrial ribosomal protein S26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030811.3 | 313 | Silent Mutation | GCA,GCG | A,A 91 | NP_110438.1 |
PTPRA - protein tyrosine phosphatase, receptor type A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |