Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607159 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZMIZ1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZMIZ1 - zinc finger MIZ-type containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020338.3 | Intron | NP_065071.1 | ||||
XM_005269987.4 | Intron | XP_005270044.1 | ||||
XM_005269988.3 | Intron | XP_005270045.1 | ||||
XM_006717923.3 | Intron | XP_006717986.1 | ||||
XM_006717924.3 | Intron | XP_006717987.1 | ||||
XM_006717925.3 | Intron | XP_006717988.1 | ||||
XM_011539978.2 | Intron | XP_011538280.1 | ||||
XM_011539979.1 | Intron | XP_011538281.1 | ||||
XM_011539980.2 | Intron | XP_011538282.1 | ||||
XM_017016440.1 | Intron | XP_016871929.1 | ||||
XM_017016441.1 | Intron | XP_016871930.1 | ||||
XM_017016442.1 | Intron | XP_016871931.1 | ||||
XM_017016443.1 | Intron | XP_016871932.1 |
ZMIZ1-AS1 - ZMIZ1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |