Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615079 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASUN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
ASUN - asunder, spermatogenesis regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018164.2 | Intron | NP_060634.2 | ||||
XM_005253422.3 | Intron | XP_005253479.1 | ||||
XM_011520750.2 | Intron | XP_011519052.1 | ||||
XM_011520751.2 | Intron | XP_011519053.1 | ||||
XM_011520752.2 | Intron | XP_011519054.1 | ||||
XM_011520753.2 | Intron | XP_011519055.1 | ||||
XM_017019632.1 | Intron | XP_016875121.1 | ||||
XM_017019633.1 | Intron | XP_016875122.1 | ||||
XM_017019634.1 | Intron | XP_016875123.1 | ||||
XM_017019635.1 | Intron | XP_016875124.1 | ||||
XM_017019636.1 | Intron | XP_016875125.1 | ||||
XM_017019637.1 | Intron | XP_016875126.1 |
Set Membership: |
HapMap Validated |