Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600144 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITPR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ITPR2 - inositol 1,4,5-trisphosphate receptor type 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002223.3 | 11368 | UTR 3 | NP_002214.2 | |||
XM_017019266.1 | 11368 | UTR 3 | XP_016874755.1 | |||
XM_017019267.1 | 11368 | Intron | XP_016874756.1 | |||
XM_017019268.1 | 11368 | Intron | XP_016874757.1 | |||
XM_017019269.1 | 11368 | Intron | XP_016874758.1 |
Set Membership: |
HapMap |