Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615836 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STK38L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
STK38L - serine/threonine kinase 38 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015000.3 | Intron | NP_055815.1 | ||||
XM_005253342.3 | Intron | XP_005253399.1 | ||||
XM_006719058.3 | Intron | XP_006719121.1 | ||||
XM_006719059.3 | Intron | XP_006719122.1 | ||||
XM_011520613.2 | Intron | XP_011518915.1 | ||||
XM_017019036.1 | Intron | XP_016874525.1 |
Set Membership: |
HapMap |