Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATCCTGCATGATTTGATCTCTAC[A/C]TCCACTTGCATCTCTTGCTGTCATC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608231 | |||||||||||||||||||||||
Literature Links: |
RASSF8 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
RASSF8 - Ras association domain family member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164746.1 | Intron | NP_001158218.1 | ||||
NM_001164747.1 | Intron | NP_001158219.1 | ||||
NM_001164748.1 | Intron | NP_001158220.1 | ||||
NM_007211.4 | Intron | NP_009142.2 | ||||
XM_006719027.2 | Intron | XP_006719090.1 | ||||
XM_006719028.1 | Intron | XP_006719091.1 | ||||
XM_017018746.1 | Intron | XP_016874235.1 |
RASSF8-AS1 - RASSF8 atnisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |