Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTAATGATACAAGTTCAACATATA[A/G]TTGAATAAAGTGAACAAGTAAACAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602892 MIM: 602893 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLRC3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KLRC3 - killer cell lectin like receptor C3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002261.2 | Intron | NP_002252.2 | ||||
NM_007333.2 | Intron | NP_031359.2 |
KLRC4 - killer cell lectin like receptor C4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLRC4-KLRK1 - KLRC4-KLRK1 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |