Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLB1L3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GLB1L3 - galactosidase beta 1 like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080407.2 | Intron | NP_001073876.2 | ||||
XM_011542566.2 | Intron | XP_011540868.1 | ||||
XM_011542567.2 | Intron | XP_011540869.1 | ||||
XM_011542568.2 | Intron | XP_011540870.1 | ||||
XM_011542569.2 | Intron | XP_011540871.1 | ||||
XM_011542570.2 | Intron | XP_011540872.1 | ||||
XM_011542571.2 | Intron | XP_011540873.1 | ||||
XM_011542572.1 | Intron | XP_011540874.1 | ||||
XM_011542573.1 | Intron | XP_011540875.1 | ||||
XM_011542574.2 | Intron | XP_011540876.1 | ||||
XM_017017155.1 | Intron | XP_016872644.1 | ||||
XM_017017156.1 | Intron | XP_016872645.1 |
Set Membership: |
HapMap |