Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 130120 MIM: 605148 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CELA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CELA1 - chymotrypsin like elastase family member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GALNT6 - polypeptide N-acetylgalactosaminyltransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007210.3 | 4746 | Intron | NP_009141.2 | |||
XM_005268607.1 | 4746 | UTR 3 | XP_005268664.1 | |||
XM_006719214.2 | 4746 | UTR 3 | XP_006719277.1 | |||
XM_011537819.2 | 4746 | UTR 3 | XP_011536121.1 | |||
XM_011537822.2 | 4746 | UTR 3 | XP_011536124.1 | |||
XM_017018744.1 | 4746 | UTR 3 | XP_016874233.1 | |||
XM_017018745.1 | 4746 | UTR 3 | XP_016874234.1 |
Set Membership: |
HapMap |