Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605148 MIM: 605024 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GALNT6 - polypeptide N-acetylgalactosaminyltransferase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC4A8 - solute carrier family 4 member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039960.2 | Intron | NP_001035049.1 | ||||
NM_001258401.2 | Intron | NP_001245330.1 | ||||
NM_001258402.1 | Intron | NP_001245331.1 | ||||
NM_001258403.1 | Intron | NP_001245332.1 | ||||
NM_001267615.1 | Intron | NP_001254544.1 | ||||
XM_006719700.1 | Intron | XP_006719763.1 | ||||
XM_011539010.1 | Intron | XP_011537312.1 | ||||
XM_011539011.1 | Intron | XP_011537313.1 | ||||
XM_011539014.2 | Intron | XP_011537316.1 | ||||
XM_017020240.1 | Intron | XP_016875729.1 | ||||
XM_017020241.1 | Intron | XP_016875730.1 | ||||
XM_017020242.1 | Intron | XP_016875731.1 |
Set Membership: |
HapMap |