Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610491 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR3665 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR3665 - microRNA 3665 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLAIN1 - SLAIN motif family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040153.3 | Intron | NP_001035243.2 | ||||
NM_001242868.1 | Intron | NP_001229797.1 | ||||
NM_001242869.1 | Intron | NP_001229798.1 | ||||
NM_001242870.1 | Intron | NP_001229799.1 | ||||
NM_001242871.1 | Intron | NP_001229800.1 | ||||
NM_144595.3 | Intron | NP_653196.1 | ||||
XM_011534929.2 | Intron | XP_011533231.1 | ||||
XM_011534930.1 | Intron | XP_011533232.1 |