Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611338 MIM: 188345 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATG4B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATG4B - autophagy related 4B cysteine peptidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DTYMK - deoxythymidylate kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165031.1 | 741 | UTR 3 | NP_001158503.1 | |||
NM_001320902.1 | 741 | UTR 3 | NP_001307831.1 | |||
NM_001320903.1 | 741 | UTR 3 | NP_001307832.1 | |||
NM_001320904.1 | 741 | UTR 3 | NP_001307833.1 | |||
NM_001320905.1 | 741 | UTR 3 | NP_001307834.1 | |||
NM_012145.3 | 741 | UTR 3 | NP_036277.2 |
Set Membership: |
Validated |