Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 113703 MIM: 602783 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPL13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RPL13 - ribosomal protein L13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000977.3 | 410 | Missense Mutation | ACC,GCC | T,A 112 | NP_000968.2 | |
NM_001243130.1 | 410 | Missense Mutation | ACC,GCC | T,A 93 | NP_001230059.1 | |
NM_001243131.1 | 410 | Intron | NP_001230060.1 | |||
NM_033251.2 | 410 | Missense Mutation | ACC,GCC | T,A 112 | NP_150254.1 |
SNORD68 - small nucleolar RNA, C/D box 68 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPG7 - SPG7, paraplegin matrix AAA peptidase subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |