Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614315 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACOT12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACOT12 - acyl-CoA thioesterase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130767.2 | Intron | NP_570123.1 | ||||
XM_006714531.2 | Intron | XP_006714594.1 | ||||
XM_006714532.3 | Intron | XP_006714595.1 | ||||
XM_017009045.1 | Intron | XP_016864534.1 | ||||
XM_017009046.1 | Intron | XP_016864535.1 | ||||
XM_017009047.1 | Intron | XP_016864536.1 | ||||
XM_017009048.1 | Intron | XP_016864537.1 |
ZCCHC9 - zinc finger CCHC-type containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131035.1 | Intron | NP_001124507.1 | ||||
NM_001131036.1 | Intron | NP_001124508.1 | ||||
NM_032280.2 | Intron | NP_115656.1 | ||||
XM_005248616.3 | Intron | XP_005248673.1 |
Set Membership: |
HapMap |