Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR5006 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR5006 - microRNA 5006 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VWA8 - von Willebrand factor A domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009814.1 | 6416 | Intron | NP_001009814.1 | |||
NM_015058.1 | 6416 | UTR 3 | NP_055873.1 | |||
XM_006719791.3 | 6416 | UTR 3 | XP_006719854.1 | |||
XM_011535007.2 | 6416 | Intron | XP_011533309.1 | |||
XM_017020469.1 | 6416 | UTR 3 | XP_016875958.1 | |||
XM_017020470.1 | 6416 | Intron | XP_016875959.1 | |||
XM_017020471.1 | 6416 | Intron | XP_016875960.1 | |||
XM_017020472.1 | 6416 | UTR 3 | XP_016875961.1 | |||
XM_017020473.1 | 6416 | UTR 3 | XP_016875962.1 | |||
XM_017020474.1 | 6416 | Intron | XP_016875963.1 |