Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608399 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSMD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CSMD3 - CUB and Sushi multiple domains 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052900.2 | Intron | NP_443132.3 | ||||
NM_198123.1 | Intron | NP_937756.1 | ||||
NM_198124.1 | Intron | NP_937757.1 | ||||
XM_011516815.2 | Intron | XP_011515117.1 | ||||
XM_011516816.2 | Intron | XP_011515118.1 | ||||
XM_017013008.1 | Intron | XP_016868497.1 | ||||
XM_017013009.1 | Intron | XP_016868498.1 | ||||
XM_017013010.1 | Intron | XP_016868499.1 | ||||
XM_017013011.1 | Intron | XP_016868500.1 | ||||
XM_017013012.1 | Intron | XP_016868501.1 |