Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607012 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BDP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BDP1 - B double prime 1, subunit of RNA polymerase III transcription initiation factor IIIB | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018429.2 | Intron | NP_060899.2 | ||||
XM_011543511.2 | Intron | XP_011541813.1 | ||||
XM_011543512.2 | Intron | XP_011541814.1 | ||||
XM_017009630.1 | Intron | XP_016865119.1 | ||||
XM_017009631.1 | Intron | XP_016865120.1 | ||||
XM_017009632.1 | Intron | XP_016865121.1 | ||||
XM_017009633.1 | Intron | XP_016865122.1 | ||||
XM_017009634.1 | Intron | XP_016865123.1 |
Set Membership: |
HapMap |