Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612441 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MGAT5B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MGAT5B - mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, isozyme B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199172.1 | Intron | NP_001186101.1 | ||||
NM_144677.2 | Intron | NP_653278.2 | ||||
NM_198955.1 | Intron | NP_945193.1 | ||||
XM_006721707.3 | Intron | XP_006721770.1 | ||||
XM_011524350.2 | Intron | XP_011522652.1 | ||||
XM_011524352.2 | Intron | XP_011522654.1 | ||||
XM_011524354.2 | Intron | XP_011522656.1 | ||||
XM_017024200.1 | Intron | XP_016879689.1 |
Set Membership: |
HapMap |