Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600967 | ||||||||||||||||||||
Literature Links: |
C8orf59 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C8orf59 - chromosome 8 open reading frame 59 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099670.2 | 1210 | UTR 3 | NP_001093140.1 | |||
NM_001099671.2 | 1210 | UTR 3 | NP_001093141.1 | |||
NM_001099672.2 | 1210 | UTR 3 | NP_001093142.1 | |||
NM_001099673.2 | 1210 | UTR 3 | NP_001093143.1 | |||
NM_001293320.1 | 1210 | UTR 3 | NP_001280249.1 | |||
XM_011517528.2 | 1210 | Intron | XP_011515830.1 |
E2F5 - E2F transcription factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083588.1 | 1210 | UTR 3 | NP_001077057.1 | |||
NM_001083589.1 | 1210 | UTR 3 | NP_001077058.1 | |||
NM_001951.3 | 1210 | UTR 3 | NP_001942.2 |