Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCTTAGTGGCCTGAGCGGCTTG[A/C]CCAGAGCTGCTGCAACTGCAGCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607328 MIM: 615695 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEXIM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HEXIM1 - hexamethylene bisacetamide inducible 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HEXIM2 - hexamethylene bisacetamide inducible 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303436.1 | 74 | Intron | NP_001290365.1 | |||
NM_001303437.1 | 74 | Intron | NP_001290366.1 | |||
NM_001303438.1 | 74 | Intron | NP_001290367.1 | |||
NM_001303439.1 | 74 | Intron | NP_001290368.1 | |||
NM_001303440.1 | 74 | UTR 5 | NP_001290369.1 | |||
NM_001303441.1 | 74 | UTR 5 | NP_001290370.1 | |||
NM_001303442.1 | 74 | UTR 5 | NP_001290371.1 | |||
NM_001303443.1 | 74 | UTR 5 | NP_001290372.1 | |||
NM_001303444.1 | 74 | UTR 5 | NP_001290373.1 | |||
NM_144608.2 | 74 | Intron | NP_653209.1 | |||
XM_006721687.3 | 74 | Intron | XP_006721750.1 | |||
XM_011524306.2 | 74 | Intron | XP_011522608.1 | |||
XM_011524307.2 | 74 | Missense Mutation | ACC,CCC | T,P 137 | XP_011522609.2 | |
XM_011524308.2 | 74 | Missense Mutation | ACC,CCC | T,P 137 | XP_011522610.2 | |
XM_017024167.1 | 74 | UTR 5 | XP_016879656.1 | |||
XM_017024168.1 | 74 | UTR 5 | XP_016879657.1 | |||
XM_017024169.1 | 74 | UTR 5 | XP_016879658.1 | |||
XM_017024170.1 | 74 | UTR 5 | XP_016879659.1 |
LOC105371795 - uncharacterized LOC105371795 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |