Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605767 | ||||||||||||||||||||
Literature Links: |
PAG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAG1 - phosphoprotein membrane anchor with glycosphingolipid microdomains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018440.3 | 7338 | UTR 3 | NP_060910.3 | |||
XM_017013640.1 | 7338 | UTR 3 | XP_016869129.1 | |||
XM_017013641.1 | 7338 | UTR 3 | XP_016869130.1 | |||
XM_017013642.1 | 7338 | UTR 3 | XP_016869131.1 | |||
XM_017013643.1 | 7338 | UTR 3 | XP_016869132.1 |