Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180630 MIM: 604983 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP95 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CEP95 - centrosomal protein 95 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DDX5 - DEAD-box helicase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320595.1 | 1840 | Missense Mutation | GCA,TCA | A,S 480 | NP_001307524.1 | |
NM_001320596.1 | 1840 | Missense Mutation | GCA,TCA | A,S 480 | NP_001307525.1 | |
NM_001320597.1 | 1840 | Missense Mutation | GCA,TCA | A,S 480 | NP_001307526.1 | |
NM_004396.4 | 1840 | Missense Mutation | GCA,TCA | A,S 480 | NP_004387.1 |
MIR3064 - microRNA 3064 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR5047 - microRNA 5047 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLG2 - DNA polymerase gamma 2, accessory subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |