Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606221 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IKZF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
IKZF3 - IKAROS family zinc finger 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257408.1 | 1157 | Silent Mutation | TCC,TCT | S,S 404 | NP_001244337.1 | |
NM_001257409.1 | 1157 | Silent Mutation | TCC,TCT | S,S 365 | NP_001244338.1 | |
NM_001257410.1 | 1157 | Silent Mutation | TCC,TCT | S,S 351 | NP_001244339.1 | |
NM_001257411.1 | 1157 | Silent Mutation | TCC,TCT | S,S 295 | NP_001244340.1 | |
NM_001257412.1 | 1157 | Silent Mutation | TCC,TCT | S,S 256 | NP_001244341.1 | |
NM_001257413.1 | 1157 | Silent Mutation | TCC,TCT | S,S 217 | NP_001244342.1 | |
NM_001257414.1 | 1157 | Silent Mutation | TCC,TCT | S,S 304 | NP_001244343.1 | |
NM_001284514.1 | 1157 | Silent Mutation | TCC,TCT | S,S 191 | NP_001271443.1 | |
NM_001284515.1 | 1157 | Silent Mutation | TCC,TCT | S,S 191 | NP_001271444.1 | |
NM_001284516.1 | 1157 | Silent Mutation | TCC,TCT | S,S 191 | NP_001271445.1 | |
NM_012481.4 | 1157 | Silent Mutation | TCC,TCT | S,S 438 | NP_036613.2 | |
NM_183228.2 | 1157 | Silent Mutation | TCC,TCT | S,S 382 | NP_899051.1 | |
NM_183229.2 | 1157 | Silent Mutation | TCC,TCT | S,S 399 | NP_899052.1 | |
NM_183230.2 | 1157 | Silent Mutation | TCC,TCT | S,S 399 | NP_899053.1 | |
NM_183231.2 | 1157 | Silent Mutation | TCC,TCT | S,S 343 | NP_899054.1 | |
NM_183232.2 | 1157 | Silent Mutation | TCC,TCT | S,S 360 | NP_899055.1 |
Set Membership: |
HapMap Validated |