Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606229 MIM: 607268 | ||||||||||||||||||||
Literature Links: |
AGO2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AGO2 - argonaute 2, RISC catalytic component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164623.1 | 12887 | Intron | NP_001158095.1 | |||
NM_012154.3 | 12887 | Intron | NP_036286.2 | |||
XM_011516965.2 | 12887 | UTR 3 | XP_011515267.1 | |||
XM_011516966.2 | 12887 | UTR 3 | XP_011515268.1 | |||
XM_011516968.2 | 12887 | UTR 3 | XP_011515270.2 | |||
XM_017013317.1 | 12887 | UTR 3 | XP_016868806.1 |
CHRAC1 - chromatin accessibility complex 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |