Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607954 MIM: 610818 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RANGRF PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RANGRF - RAN guanine nucleotide release factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177801.1 | 679 | UTR 3 | NP_001171272.1 | |||
NM_001177802.1 | 679 | UTR 3 | NP_001171273.1 | |||
NM_016492.4 | 679 | Intron | NP_057576.2 | |||
XM_005256618.4 | 679 | Intron | XP_005256675.1 |
SLC25A35 - solute carrier family 25 member 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320870.1 | 679 | Intron | NP_001307799.1 | |||
NM_001320871.1 | 679 | UTR 3 | NP_001307800.1 | |||
NM_001320872.1 | 679 | UTR 3 | NP_001307801.1 | |||
NM_001320877.1 | 679 | Intron | NP_001307806.1 | |||
NM_201520.2 | 679 | UTR 3 | NP_958928.1 | |||
XM_005256641.2 | 679 | Intron | XP_005256698.1 | |||
XM_011523846.1 | 679 | UTR 3 | XP_011522148.1 | |||
XM_017024633.1 | 679 | Intron | XP_016880122.1 | |||
XM_017024634.1 | 679 | Intron | XP_016880123.1 |