Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTATTTTCCCACCTGTTTGCTGTA[A/C]GATGACTATTATAATCACTCGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
59 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616574 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105378614 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105378614 - uncharacterized LOC105378614 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MINOS1 - mitochondrial inner membrane organizing system 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032363.3 | Intron | NP_001027535.1 | ||||
NM_001204082.1 | Intron | NP_001191011.1 | ||||
NM_001204083.1 | Intron | NP_001191012.1 |
MINOS1-NBL1 - MINOS1-NBL1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204088.1 | Intron | NP_001191017.1 | ||||
NM_001204089.1 | Intron | NP_001191018.1 |
RPS14P3 - ribosomal protein S14 pseudogene 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |