Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC178 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC178 - coiled-coil domain containing 178 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105528.1 | Intron | NP_001098998.1 | ||||
NM_001308126.1 | Intron | NP_001295055.1 | ||||
NM_198995.2 | Intron | NP_945346.2 | ||||
XM_011525948.1 | Intron | XP_011524250.1 | ||||
XM_011525951.1 | Intron | XP_011524253.1 | ||||
XM_011525954.2 | Intron | XP_011524256.1 | ||||
XM_011525955.1 | Intron | XP_011524257.1 | ||||
XM_017025721.1 | Intron | XP_016881210.1 | ||||
XM_017025722.1 | Intron | XP_016881211.1 | ||||
XM_017025723.1 | Intron | XP_016881212.1 | ||||
XM_017025724.1 | Intron | XP_016881213.1 | ||||
XM_017025725.1 | Intron | XP_016881214.1 |