Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609150 MIM: 156535 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXXC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CXXC1 - CXXC finger protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101654.1 | 2472 | UTR 3 | NP_001095124.1 | |||
NM_014593.3 | 2472 | UTR 3 | NP_055408.2 | |||
XM_011525940.2 | 2472 | UTR 3 | XP_011524242.1 | |||
XM_011525941.2 | 2472 | UTR 3 | XP_011524243.1 | |||
XM_017025718.1 | 2472 | UTR 3 | XP_016881207.1 | |||
XM_017025719.1 | 2472 | UTR 3 | XP_016881208.1 |