Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603684 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPG PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LIPG - lipase G, endothelial type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308006.1 | 290 | Silent Mutation | TCA,TCC | S,S 4 | NP_001294935.1 | |
NM_006033.3 | 290 | Silent Mutation | TCA,TCC | S,S 4 | NP_006024.1 | |
XM_005258390.1 | 290 | Intron | XP_005258447.1 | |||
XM_011526265.2 | 290 | Intron | XP_011524567.1 | |||
XM_011526267.1 | 290 | Intron | XP_011524569.1 | |||
XM_017026095.1 | 290 | Intron | XP_016881584.1 |
Set Membership: |
HapMap |