Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616054 MIM: 608731 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ELP2 - elongator acetyltransferase complex subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242875.2 | Intron | NP_001229804.1 | ||||
NM_001242876.2 | Intron | NP_001229805.1 | ||||
NM_001242877.2 | Intron | NP_001229806.1 | ||||
NM_001242878.2 | Intron | NP_001229807.1 | ||||
NM_001242879.2 | Intron | NP_001229808.1 | ||||
NM_001324465.1 | Intron | NP_001311394.1 | ||||
NM_001324466.1 | Intron | NP_001311395.1 | ||||
NM_001324467.1 | Intron | NP_001311396.1 | ||||
NM_001324468.1 | Intron | NP_001311397.1 | ||||
NM_018255.3 | Intron | NP_060725.1 |
SLC39A6 - solute carrier family 39 member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |