Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTTGGGGGAACACACAGAAATGG[A/G]TCGGAACACAATGGAGAGAGACGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607875 MIM: 600174 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INPP5K PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
INPP5K - inositol polyphosphate-5-phosphatase K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PITPNA - phosphatidylinositol transfer protein alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006224.3 | 1358 | UTR 3 | NP_006215.1 | |||
XM_011523939.1 | 1358 | UTR 3 | XP_011522241.1 | |||
XM_011523940.1 | 1358 | UTR 3 | XP_011522242.1 |
PITPNA-AS1 - PITPNA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |