Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612077 MIM: 614218 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR22 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR22 - microRNA 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR22HG - MIR22 host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR81 - WD repeat domain 81 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163673.1 | Intron | NP_001157145.1 | ||||
NM_001163809.1 | Intron | NP_001157281.1 | ||||
NM_001163811.1 | Intron | NP_001157283.1 | ||||
NM_152348.3 | Intron | NP_689561.2 | ||||
XM_011523651.2 | Intron | XP_011521953.1 | ||||
XM_017024184.1 | Intron | XP_016879673.1 |