Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607145 MIM: 601594 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DTNBP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DTNBP1 - dystrobrevin binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271667.1 | 1350 | UTR 3 | NP_001258596.1 | |||
NM_001271668.1 | 1350 | UTR 3 | NP_001258597.1 | |||
NM_001271669.1 | 1350 | UTR 3 | NP_001258598.1 | |||
NM_032122.4 | 1350 | UTR 3 | NP_115498.2 | |||
NM_183040.2 | 1350 | Intron | NP_898861.1 | |||
XM_005249447.4 | 1350 | UTR 3 | XP_005249504.1 | |||
XM_011514936.2 | 1350 | UTR 3 | XP_011513238.1 | |||
XM_011514937.2 | 1350 | UTR 3 | XP_011513239.1 | |||
XM_017011348.1 | 1350 | UTR 3 | XP_016866837.1 | |||
XM_017011349.1 | 1350 | UTR 3 | XP_016866838.1 |
JARID2 - jumonji and AT-rich interaction domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267040.1 | 1350 | Intron | NP_001253969.1 | |||
NM_004973.3 | 1350 | Intron | NP_004964.2 | |||
XM_005249089.3 | 1350 | Intron | XP_005249146.1 | |||
XM_011514579.2 | 1350 | Intron | XP_011512881.1 | |||
XM_011514584.2 | 1350 | Intron | XP_011512886.1 | |||
XM_017010833.1 | 1350 | Intron | XP_016866322.1 | |||
XM_017010834.1 | 1350 | Intron | XP_016866323.1 | |||
XM_017010835.1 | 1350 | Intron | XP_016866324.1 |
Set Membership: |
HapMap |